missing translation for 'onlineSavingsMsg'
Learn More

Cobalt slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.95% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol

$181.82 - $2256.05

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4235406
View Documents
Thermo Scientific Chemicals
04235406
5 g
Each for $181.82
Only null left
 
AA4235414
View Documents
Thermo Scientific Chemicals
04235414
25 g
Each for $692.00
Only null left
 
AA4235422
View Documents
Thermo Scientific Chemicals
04235422
100 g
Each for $2,256.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-48-4
58.93
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
CHEBI:27638
[Co]

Specifications

7440-48-4
Odorless
Slug
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
104730
99.95%
(metals basis)
5 g
≈0.45g/slug
Co
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
[Co]
58.93
CHEBI:27638
Cobalt slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H350

EINECSNumber : 231-158-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only