missing translation for 'onlineSavingsMsg'
Learn More

Cobalt powder, spherical, -100+325 mesh, 99.8% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol

$192.42 - $688.47

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4160518
View Documents
Thermo Scientific Chemicals
04160518
50 g
Each for $192.42
Only null left
 
AA4160530
View Documents
Thermo Scientific Chemicals
04160530
250 g
Each for $688.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-48-4
58.93
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
CHEBI:27638
[Co]

Specifications

Black
50 g
Odorless
Co
UN3089
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
104730
99.8%
(metals basis)
7440-48-4
−100+325 Mesh
Spherical Powder
MFCD00010935
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
[Co]
58.93
CHEBI:27638
Cobalt powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H334-H317
Flammable solid.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P201-P202-P210-P240-P241-P260-P264b-P270-P271-P272-P280-P281-P284-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P378q-P501c

H228-H302-H317-H319-H330-H334-H335-H336-H350i-H360-H373-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 231-158-0

RTECSNumber : GF8750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only