missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Nitrate Hexahydrate, Honeywell Fluka™

ACS Reagent,≥98%

$157.40 - $910.20

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name cobalt(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000424
View Documents
Honeywell Chemicals
2392675G
5 g
Each for $157.40
Only null left
 
6000422
View Documents
Honeywell Chemicals
239267100G
100 g
Each for $233.60
Only null left
 
6000423
View Documents
Honeywell-Fluka
239267500G
500 g
Each for $910.20
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10026-22-9
291.03
QGUAJWGNOXCYJF-UHFFFAOYSA-N
24821
cobalt(2+);dinitrate;hexahydrate
CoH12N2O12
MFCD00149647
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
CHEBI:86214
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

10026-22-9
CoH12N2O12
MFCD00149647
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
291.03
CHEBI:86214
≥98%
Cobalt(Ii) Nitrate Hexahydrate
5 g
Co(NO3)2 · 6H2O
1477
QGUAJWGNOXCYJF-UHFFFAOYSA-N
cobalt(2+);dinitrate;hexahydrate
24821
291.03g/mol
ACS Reagent
Videos
Safety and Handling

Safety and Handling

P201-P210-P261-P280-P284-P304 + P340-P308 + P313

H272-H302-H317-H334-H341-H350i-H360F-H410

RTECSNumber : QU7355500

SDS
missing translation for 'documents'

missing translation for 'documents'