Learn More
Cobalt foil, 0.5mm (0.02in) thick, 99.95% (metals basis)
CAS: 7440-48-4 | Co | 58.93 g/mol
$148.29 - $1223.36
Chemical Identifiers
CAS | 7440-48-4 |
---|---|
Molecular Formula | Co |
Molecular Weight (g/mol) | 58.93 |
MDL Number | MFCD00010935 |
InChI Key | GUTLYIVDDKVIGB-UHFFFAOYSA-N |
Synonym | kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride |
PubChem CID | 104730 |
ChEBI | CHEBI:27638 |
IUPAC Name | cobalt |
SMILES | [Co] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA42659FF
|
Thermo Scientific Chemicals
042659FF |
25 x 25 mm |
Each for $148.29
|
|
|||||
AA42659FI
|
Thermo Scientific Chemicals
042659FI |
50 x 50 mm |
Each for $427.21
|
|
|||||
AA42659GH
|
Thermo Scientific Chemicals
042659GH |
100 x 100 mm |
Each for $1,223.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-48-4 | |
58.93 | |
GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
104730 | |
cobalt |
Co | |
MFCD00010935 | |
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
CHEBI:27638 | |
[Co] |
Specifications
7440-48-4 | |
Odorless | |
Foil | |
Co | |
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
[Co] | |
58.93 | |
CHEBI:27638 | |
Cobalt foil, 0.5mm (0.02 in.) thick |
25 x 25 mm | |
≈2.8g/25x25mm | |
99.95% (metals basis) | |
MFCD00010935 | |
GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
cobalt | |
104730 | |
99.95% | |
(metals basis) |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350
EINECSNumber : 231-158-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only