missing translation for 'onlineSavingsMsg'
Learn More

Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL

$86.86 - $165.77

Chemical Identifiers

MDL Number MFCD00010935
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA88060AE
View Documents
Thermo Scientific Chemicals
088060AE
100 mL
Each for $86.86
Only null left
 
AA88060AP
View Documents
Thermo Scientific Chemicals
088060AP
500 mL
Each for $165.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00010935

Specifications

Liquid
MFCD00010935
100 mL
Cobalt, AAS standard solution, Co 1000μg/mL
Co in 5% HNO3
UN3264
Specpure™
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335-H350i-H360F

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only