Learn More
Brady™ Clear Acrylic Overlaminate for BradyJet Labels
Achieve up to 10 years of outdoor durability through improved UV and abrasion resistance with acrylic overlaminate for BradyJet labels.
Supplier:  Brady™ LAM261910
 View more versions of this product
Catalog No. 17-377-018
Only null left
Explore available promotions
Description
Description
  • No need to cut to size
  • Simply peel and apply with these pre-sized rolls ideally sized to their corresponding BradyJet labels
  • Overlaminate is a quarter-inch wider than its corresponding BradyJet label to ensure complete coverage
Specifications
Specifications
Not Autoclavable
None
Gloss
Acrylic
Ink Jet
Non-sterile
-29°C
81.28 x 116.33 mm
-28.9°C to +82.2°C
0.076 mm
3.2 in.
4.58 in.
Permanent Acrylic
Clear
Roll
Single-side Printable
Labels
Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Pipe and Valve Marking, Warehouse Marking
270 Labels
-20°F to +180°F
0.003 in.
3.2 x 4.58 in.
81.28 mm
116.33 mm
Product Suggestions
Product Suggestions