missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Clear Acrylic Overlaminate for BradyJet Labels
Achieve up to 10 years of outdoor durability through improved UV and abrasion resistance with acrylic overlaminate for BradyJet labels.
Supplier: Brady™ LAM262910
Description
- No need to cut to size
- Simply peel and apply with these pre-sized rolls ideally sized to their corresponding BradyJet labels
- Overlaminate is a quarter-inch wider than its corresponding BradyJet label to ensure complete coverage
Specifications
Not Autoclavable | |
None | |
Gloss | |
Acrylic | |
Ink Jet | |
Non-sterile | |
-29°C | |
111.25 x 161.29 mm | |
-28.9°C to +82.2°C | |
0.076 mm | |
4.38 in. | |
6.35 in. |
Permanent Acrylic | |
Clear | |
Roll | |
Single-side Printable | |
Labels | |
Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Pipe and Valve Marking, Warehouse Marking | |
200 Labels | |
-20°F to +180°F | |
0.003 in. | |
4.38 x 6.35 in. | |
111.25 mm | |
161.29 mm |