Learn More
Agilent Tech Canada Inc CL BTO INLET SEPTA 9MM 50PK 58 DFS Item
Supplier:  Agilent Tech Canada Inc 80100217
CrossLab Non-stick BTO inlet septa 9mm CenterGuide 50/PK
Catalog No. NC0527139
Only null left
Explore available promotions