missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Monohydrate, Crystalline Powder, USP, 99.5-100.5%, Spectrum™ Chemical

C1296, 5949-29-1, C6H8O7·H2O

$611.40 - $7844.10

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601721
View Documents
Spectrum Chemical Mfg Cor
C1296500GM
500 g Poly Bottle
Each for $611.40
Only null left
 
18601722
View Documents
Spectrum Chemical Mfg Cor
C129625KG
2.5 kg Poly Bottle
Each for $1,820.97
Only null left
 
18601723
View Documents
Spectrum Chemical Mfg Cor
C129612KG
12 kg Poly Pail
Each for $3,868.53
Only null left
 
18601724
View Documents
Spectrum Chemical Mfg Cor
C129650KGBL
50 kg Fiber Drum
Each for $7,844.10
Only null left
 
Description

Description

Spectrum™ Chemical Citric Acid, Monohydrate, Crystalline Powder, USP is used as thrombin inhibitor and fungicide in pharmaceutical prepartions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

5949-29-1
210.14
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
C6H10O8
YASYEJJMZJALEJ-UHFFFAOYSA-N
O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

5949-29-1
C6H10O8
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
210.14
USP
0.001
1
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
99.5 to 100.5%
500 g
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'