missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Anhydrous, ACS, 99.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C1280125GM
Specifications
77-92-9 | |
99.5% | |
125 g | |
0.0002 |
1 | |
ACS | |
Poly Bottle |
Chemical Identifiers
77-92-9 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.