missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, ≥99.5%, ACS reagent, anhydrous

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol

$140.38 - $381.17

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC423565000
SDS View Documents
Thermo Scientific Chemicals
423565000
500 g
Each for $140.38
Case of 6 Each for $842.28
Only null left
 
AC423560020
SDS View Documents
Thermo Scientific Chemicals
423560020
2 kg
Each for $381.17
Case of 6 Each for $2,287.02
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

77-92-9
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
311
2-hydroxypropane-1,2,3-tricarboxylic acid
C6H8O7
MFCD00011669
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
CHEBI:30769
OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

153.0°C
C6H8O7
MFCD00011669
Solubility in water: 750g/L (20°C). Other solubilities: soluble in alcohol and ether, badly soluble in diethyl ether, insoluble in benzene and chloroform
OC(=O)CC(O)(CC(O)=O)C(O)=O
192.12
CHEBI:30769
99.6%
500 g
Citric acid
77-92-9
HO2CCH2C(OH)(CO2H)CH2CO2H
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
311
192.13
ACS Reagent
100°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only