missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, 10% w/v, Reagents
Citric Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$200.03 - $480.30
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | 2-hydroxypropane-1,2,3-tricarboxylic Acid |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-335-603
|
Labchem Inc
CC2041001A |
1 L |
Each for $200.03
|
|
|||||
01-335-602
|
Labchem Inc
CC2041004A |
4 L |
Each for $480.30
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
2-hydroxypropane-1,2,3-tricarboxylic Acid | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
1.72 | |
101°C | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
10% w/v | |
1 L | |
Liquid | |
Citric Acid, 10% w/v |
7732-18-5,77-92-9 | |
Colorless | |
2-hydroxypropane-1,2,3-tricarboxylic Acid | |
O | |
18.02 | |
Reagent | |
Poly Bottle | |
1.0405 |