Learn More
Mopec™ Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats
Available in multiple sizes and colors for use with Leica, TBS, and Tanner Cryostats.
Supplier:  Mopec™ SP101RD
 View more versions of this product
Catalog No. 22-445-320
Only null left
Explore available promotions
Description
Description
  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats
Specifications
Specifications
Leica, TBS, and Tanner Cryostats
22 mm
Red
Product Suggestions
Product Suggestions