missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium Acetate, Basic, Powder, Purified, Spectrum™ Chemical
$416.27 - $7462.87
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18609744
|
Spectrum Chemical Mfg Cor
C1236125GM |
125 g | Amber Glass Bottle |
Each for $416.27
|
|
||||
18609745
|
Spectrum Chemical Mfg Cor
C1236500GM |
Each for $893.60
|
|
||||||
18609746
|
Spectrum Chemical Mfg Cor
C123625KG |
2.5 kg | Poly Pail |
Each for $3,375.70
|
|
||||
18609747
|
Spectrum Chemical Mfg Cor
C123612KG |
12 kg | Poly Pail |
Each for $4,328.43
|
|
||||
18609748
|
Spectrum Chemical Mfg Cor
C123645KGBL |
45 kg | Fiber Drum |
Each for $7,462.87
|
|
||||
Chemical Identifiers
39430-51-8 |
Specifications
39430-51-8 | |
Amber Glass Bottle | |
Purified |
1 | |
125 g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.