missing translation for 'onlineSavingsMsg'
Learn More

Chlorophenol Red, 0.04% Aqueous, pH 5.2 to 6.8 Yellow to Red, Certified, LabChem™

$52.23 - $116.10

Chemical Identifiers

CAS 123333-64-2
Molecular Formula C19H11Cl2O5S
Molecular Weight (g/mol) 422.25
MDL Number MFCD00151199
InChI Key QXTPRQZMDKBTAI-UNOMPAQXSA-M
PubChem CID 131846007
IUPAC Name 2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
SMILES OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC130807
View Documents
LabChem
LC130807
125 mL
Each for $52.23
Only null left
 
LC130802
View Documents
LabChem
LC130802
1 L
Each for $116.10
Only null left
 
Description

Description

Specifications

Chemical Identifiers

123333-64-2
422.25
QXTPRQZMDKBTAI-UNOMPAQXSA-M
2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
C19H11Cl2O5S
MFCD00151199
131846007
OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O

Specifications

123333-64-2,7732-18-5
C19H11Cl2O5S
MFCD00151199
Passes Test
OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O
422.25
445.25
Amber Glass
Red
Liquid
99.96,0.04
C19H11Cl2O5SNa
Soluble in water
QXTPRQZMDKBTAI-UNOMPAQXSA-M
2-[(3-chloro-4-hydroxyphenyl)[(1Z)-3-chloro-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
131846007
Certified
Hydrogen chloride; Carbon monoxide; Carbon dioxide
125 mL
Chlorophenol Red, 0.04% Aqueous, pH 5.2 to 6.8 Yellow to Red
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'