missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Purified, Reagents
Chloroform, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
$154.87 - $2071.50
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | Trichloromethane, Methyl trichloride |
IUPAC Name | trichloromethane |
SMILES | ClC(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-337-323
|
Labchem Inc
C21755001C |
1 L |
Each for $255.53
|
|
|||||
01-337-322
|
Labchem Inc
C217550025C |
2.5 L |
Each for $298.50
|
|
|||||
01-337-321
|
Labchem Inc
C217550020E6 |
20 L |
Each for $2,071.50
|
|
|||||
01-337-320
|
Labchem Inc
C21755004C |
4 L |
Each for $477.63
|
|
|||||
01-337-319
|
Labchem Inc
C2175500500C |
500 mL |
Each for $154.87
|
|
|||||
Chemical Identifiers
67-66-3 | |
119.37 | |
Trichloromethane, Methyl trichloride | |
ClC(Cl)Cl |
CHCl3 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane |
Specifications
-63.5°C | |
Colorless | |
1 L | |
Liquid | |
CHCl3 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
Pure |
67-66-3 | |
60.5°C to 61.2°C | |
Glass Bottle | |
1.484 | |
Trichloromethane, Methyl trichloride | |
ClC(Cl)Cl | |
119.37 | |
Chloroform, Purified |