missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, puriss. p.a., ≥99.5% (GC), ACS Reagent, Honeywell Riedel-de Haën™

Puriss. p.a., ACS Reagent,≥99.5% (GC)

$281.50 - $478.55

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045434
View Documents
Honeywell Chemicals
235701L
1 L Glass Bottle
Each for $281.50
Only null left
 
60045435
View Documents
Honeywell-Riedel-de Haen
2357025L
2.5 L Glass Bottle
Each for $478.55
Only null left
 
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

108-90-7
112.556
MVPPADPHJFYWMZ-UHFFFAOYSA-N
7964
chlorobenzene
C6H5Cl
MFCD00000530
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
CHEBI:28097
C1=CC=C(C=C1)Cl

Specifications

-45°C
1.106g/mL at 25°C
1 L
27°C (80.6°F)
Glass Bottle
n20/D 1.524(lit.)
MFCD00000530
UN1134
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
C1=CC=C(C=C1)Cl
112.556
CHEBI:28097
≥99.5% (GC)
Chlorobenzene
108-90-7
132°C
0.071
1178°F
Liquid
C6H5Cl
11.8mmHg (25°C)
605632
MVPPADPHJFYWMZ-UHFFFAOYSA-N
chlorobenzene
7964
112.56g/mol
ACS/puriss. p.a.
Videos
Safety and Handling

Safety and Handling

P210-P261-P370 + P378

H226-H315-H332-H411

DOTInformation : Class 3 - PG 3 - Chlorobenzene

EINECSNumber : 203-628-5

RTECSNumber : CZ0175000

SDS
missing translation for 'documents'

missing translation for 'documents'