missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroacetic Acid, Crystal, ACS, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor M132025KG
Specifications
79-11-8 | |
0.0002 | |
C2H3ClO2 | |
FOCAUTSVDIKZOP-UHFFFAOYSA-N | |
2-chloroacetic acid | |
0.99 | |
0.0001 |
1 | |
Amber Glass Bottle | |
2.5 kg | |
OC(=O)CCl | |
94.49 | |
ACS |
Chemical Identifiers
79-11-8 | |
94.49 | |
2-chloroacetic acid |
C2H3ClO2 | |
FOCAUTSVDIKZOP-UHFFFAOYSA-N | |
OC(=O)CCl |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.