Learn More
National Safety Apparel™ Chemical Resistant FR Lab Coats, No Pockets
Chemical Resistant Lab Coat made of flame-resistant CR/FR fabric for multi-hazard protection without weighing you down.
$595.92 - $655.53
Specifications
Product Type Chemical Resistant Lab Coat
Certifications/Compliance ASTM F1506, NFPA 2112
View More Specs
Products 5
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
17000306
View Documents
National Safety Apparel™
LABCRNPNBXS
Each for $595.92
Only null left
17000307
View Documents
National Safety Apparel™
LABCRNPNBSM
Each for $595.92
Only null left
17000309
View Documents
National Safety Apparel™
LABCRNPNBLG
Each for $595.92
Only null left
17000310
View Documents
National Safety Apparel™
LABCRNPNBXL
Each for $595.92
Only null left
17000311
View Documents
National Safety Apparel™
LABCRNPNB2X
Each for $655.53
Only null left
Description
Description
  • Fabric/Material 4.4 oz. CR/FR Aramid Blend FR Nomex knit wrists keep sleeves in place
  • Bottom side vents for increased mobility
  • Durable hi-temp plastic snaps for easy donning and doffing
  • Side pass-through allows access to pockets underneath the coat
  • Durable accessory loop for safety glasses, pens, badges, etc.
  • Highly breathable
  • Easy care and laundering
Specifications
Specifications
Chemical Resistant Lab Coat
ASTM F1506, NFPA 2112
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'