missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ Chemical Resistant FR Lab Coats with Pockets
Navy chemical resistant lab coat made of flame-resistant CR/FR fabric for multi-hazard protection. Pockets feature covered flaps to secure contents.
$647.10 - $711.83
Specifications
Product Type | Chemical Resistant Lab Coat |
---|---|
Gender | Unisex |
Closure Type | Snaps |
Resistance | Flame |
Certifications/Compliance | ASTM F1506, NFPA 2112 |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17000300
|
National Safety Apparel™
LABCRNBXS |
Each for $647.10
|
|
||||||
17000301
|
National Safety Apparel™
LABCRNBSM |
Small |
Each for $647.10
|
|
|||||
17000302
|
National Safety Apparel™
LABCRNBMD |
Each for $647.10
|
|
||||||
17000303
|
National Safety Apparel™
LABCRNBLG |
Each for $647.10
|
|
||||||
17000304
|
National Safety Apparel™
LABCRNBXL |
X-Large |
Each for $647.10
|
|
|||||
17000305
|
National Safety Apparel™
LABCRNB2X |
2X-Large |
Each for $711.83
|
|
|||||
Description
- Fabric/Material 4.4 oz. CR/FR Aramid Blend FR Nomex knit wrists keep sleeves in place
- Bottom side vents for increased mobility
- Durable hi-temp plastic snaps for easy donning and doffing
- Side pass-through allows access to pockets underneath the coat
- Durable accessory loop for safety glasses, pens, badges, etc.
- Chest pocket with covered flap and pen/pencil slot
- Lower pockets with covered flaps to secure contents
- Highly breathable
- Easy care and laundering
Specifications
Chemical Resistant Lab Coat | |
Snaps | |
ASTM F1506, NFPA 2112 |
Unisex | |
Flame |