missing translation for 'onlineSavingsMsg'
Learn More

CHD4 Recombinant Human Monoclonal Antibody (K49015_15E2), Invitrogen™

Human Recombinant Monoclonal Antibody

$1421.98 - $2134.69

Specifications

Antigen CHD4
Clone K49015_15E2
Concentration 1 mg/mL
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA555994
View Documents
Invitrogen™
MA555994
200 μg
Each for $2,134.69
Only null left
 
MA555612
View Documents
Invitrogen™
MA555612
50 μg
Each for $1,421.98
Only null left
 
Description

Description

Product is compatible with indirect ELISA (1 μg/mL, capture antibody). Sequence of this protein is as follows: EMGEEEEVER EIIKQEESVD PDYWEKLLRH HYEQQQEDLA RNLGKGKRIR KQVNYNDGSQ EDRDWQDDQS DNQSDYSVAS EEGDEDFDER SEAPRRPSRK GLRNDKDKPL PPLLARVGGN IEVLGFNARQ RKAFLNAIMR YGMPPQDAFT TQWLVRDLRG KSEKEFKAYV SLFMRHLCEP GADGAETFAD GVPREGLSRQ HVLTRIGVMS LIRKKVQEFE HVNGRWSMPE LAEVEENKKM SQPGSPSPKT PTPSTPGDTQ

CHD4 Recombinant Monoclonal antibody specifically detects CHD4 in Human samples. It is validated for ELISA, Western Blot
Specifications

Specifications

CHD4
1 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide
9530019N15Rik; AA617397; ATP-dependent helicase CHD4; BC005710; Chd4; CHD-4; chromodomain helicase DNA binding protein 4; chromodomain-helicase-DNA-binding protein 4; D6Ertd380e; Mi-2; Mi-2 autoantigen; Mi-2 autoantigen 218 kDa protein; Mi-2 beta; MI2B; Mi-2b; Mi-2beta; Mi2-BETA; mKIAA4075
Recombinant human CHD4 (immunogen range is 1291-1550).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
CHD4
K49015_15E2
ELISA, Western Blot
Unconjugated
Human
Human
Q14839
1108
IgG1
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'