missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Wood, Powder, Spectrum™ Chemical
$283.76 - $14549.79
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18609679
|
Spectrum Chemical Mfg Cor
C1217500GM |
500 g | Amber Glass Bottle |
Each for $283.76
|
|
||||
18609677
|
Spectrum Chemical Mfg Cor
C121725KG |
2.5 kg | Poly Pail |
Each for $932.18
|
|
||||
18609676
|
Spectrum Chemical Mfg Cor
C121712KGBL |
12 kg | Fiber Drum |
Each for $5,129.18
|
|
||||
18609678
|
Spectrum Chemical Mfg Cor
C121750KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7440-44-0 |
Specifications
7440-44-0 | |
Amber Glass Bottle | |
Reagent |
1 | |
500 g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.