missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Activated, Powder, Spectrum™ Chemical
$350.50 - $17080.62
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18609672
|
Spectrum Chemical Mfg Cor
CH103500GM |
500 g | Amber Glass Bottle |
Each for $350.50
|
|
||||
18609670
|
Spectrum Chemical Mfg Cor
CH10340LBBL |
40 lb. | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18609671
|
Spectrum Chemical Mfg Cor
CH10345KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7440-44-0 |
Specifications
7440-44-0 | |
Amber Glass Bottle | |
Reagent |
1 | |
500 g |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.