missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Activated, Norit(R) CA3, Powder, Spectrum™ Chemical
$313.12 - $5412.85
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.01 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
IUPAC Name | carbon |
SMILES | [C] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18609659
|
Spectrum Chemical Mfg Cor
C1191500GM |
500 g | Amber Glass Bottle |
Each for $313.12
|
|
||||
18609660
|
Spectrum Chemical Mfg Cor
C119125KG |
2.5 kg | Poly Pail |
Each for $1,052.00
|
|
||||
18609661
|
Spectrum Chemical Mfg Cor
C119120KGBL |
20 kg | Fiber Drum |
Each for $5,412.85
|
|
||||
Chemical Identifiers
7440-44-0 | |
12.01 | |
carbon |
C | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
[C] |
Specifications
7440-44-0 | |
2.0 to 3.5 | |
C | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
Reagent |
1 | |
Amber Glass Bottle | |
500 g | |
[C] | |
12.01 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.