missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, NanoArc™ CE-0440, 25% in H{2}O, colloidal dispersion

Suitable for use in ultrafine acid side polishing slurries | CAS: 1306-38-3 | CeO2 | 172.114 g/mol

$103.79 - $391.93

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4676322
View Documents
Thermo Scientific Chemicals
04676322
100 g
Each for $103.79
Only null left
 
AA4676336
View Documents
Thermo Scientific Chemicals
04676336
500 g
Each for $391.93
Only null left
 
Description

Description

Cerium(IV) is suitable for use in ultrafine acid side polishing slurries.Cerium(IV) oxide is widely applied as a solid electrolyte in fuel cells. It is also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass. It is utilized to increase specific surface area and inhibit sintering as well as pore size growth at a high temperature by addition with CuO/TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1306-38-3
172.114
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CHEBI:79089
O=[Ce]=O

Specifications

1306-38-3
100 g
MFCD00010933
Immiscible with water.
O=[Ce]=O
172.114
CHEBI:79089
25% in H2O
Cerium (IV) oxide, NanoArc™ CE-6440, Colloidal dispersion
Powder
CeO2
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
Odorless
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-150-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only