Learn More
Cephalothin sodium salt
A first-generation cephalosporin antibiotic | CAS: 58-71-9 | C16H15N2NaO6S2 | 418.414 g/mol
$187.92 - $646.07
Chemical Identifiers
CAS | 58-71-9 |
---|---|
Molecular Formula | C16H15N2NaO6S2 |
Molecular Weight (g/mol) | 418.414 |
MDL Number | MFCD00072025 |
InChI Key | VUFGUVLLDPOSBC-XRZFDKQNSA-M |
Synonym | 7-(2-Thienylacetamido)cephalosporanic acid sodium salt; Cephalotin sodium salt |
PubChem CID | 23675321 |
ChEBI | CHEBI:3542 |
IUPAC Name | sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
SMILES | CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6560703
|
Thermo Scientific Chemicals
J6560703 |
1 g |
Each for $187.92
|
|
|||||
AAJ6560706
|
Thermo Scientific Chemicals
J6560706 |
5 g |
Each for $646.07
|
|
|||||
Description
Cephalothin sodium salt is used to study the mechanism of liposome encapsulated antibiotics1, strategies for co-opting β-lactamases of Gram-negative bacteria for treatment of antibiotics2, and for immunology studies in relation to antibiotics.3 It is used to study the effect of expression, binding and inhibition of penicillin-binding proteins especially PBP6 on bacterial cell wall mucopeptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCephalothin sodium salt is used to study the mechanism of liposome encapsulated antibiotics1, strategies for co-opting β-lactamases of Gram-negative bacteria for treatment of antibiotics2, and for immunology studies in relation to antibiotics.3 It is used to study the effect of expression, binding and inhibition of penicillin-binding proteins especially PBP6 on bacterial cell wall mucopeptide synthesis.
Solubility
Soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
58-71-9 | |
418.414 | |
VUFGUVLLDPOSBC-XRZFDKQNSA-M | |
23675321 | |
sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
C16H15N2NaO6S2 | |
MFCD00072025 | |
7-(2-Thienylacetamido)cephalosporanic acid sodium salt; Cephalotin sodium salt | |
CHEBI:3542 | |
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+] |
Specifications
Cephalothin Sodium Salt | |
58-71-9 | |
White | |
C16H15N2NaO6S2 | |
4120706 | |
7-(2-Thienylacetamido)cephalosporanic acid sodium salt; Cephalotin sodium salt | |
VUFGUVLLDPOSBC-XRZFDKQNSA-M | |
sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
23675321 | |
418.42 |
Powder | |
Odorless | |
1 g | |
MFCD00072025 | |
14,1982 | |
Soluble in water | |
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+] | |
418.414 | |
CHEBI:3542 | |
Cephalothin sodium salt |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
EINECSNumber : 200-394-6
RTECSNumber : XI0388300
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only