missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLMASTER Polyethylene Terephthalate (PET) Roller Bottles - Short Form
For use in cell culture. Greiner Bio-One CELLMASTER Polyethylene Terephthalate (PET) Roller Bottles - Short Form is suitable for many adherent cell lines, is stable and gas permeable.
Supplier: Greiner Bio-One 680170
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic, non-cytotoxic
- Seamless blow-moulding technique, rules out leaking seams
- USP Class VI end product testing certified
- Safety screw cap guarantees tightly closed, co
Specifications
Transparent | |
PET | |
30 | |
850cm2 | |
Free of detectable DNase/RNase, human DNA and pyrogens, Contents non-cytotoxic. | |
Yes |
277 x 116 x 51mm | |
30 | |
Bottle: -60°C to +60°C Screw cap: -50°C to +100°C | |
Blue | |
Not Autoclavable | |
116 mm |