missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD95 (APO-1/Fas) Monoclonal Antibody (DX2), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$604.08 - $1340.54
Specifications
Antigen | CD95 (APO-1/Fas) |
---|---|
Clone | DX2 |
Concentration | 0.4 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H084T03R03A
|
Invitrogen™
H084T03R03A |
100 Tests |
Each for $1,340.54
|
|
|||||
H084T02R03A
|
Invitrogen™
H084T02R03A |
25 Tests |
Each for $604.08
|
|
|||||
Description
CD95 (APO-1/Fas) Monoclonal antibody specifically detects CD95 (APO-1/Fas) in Canine, Human samples. It is validated for Flow CytometrySpecifications
CD95 (APO-1/Fas) | |
0.4 μg/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
AI196731; ALPS1A; APO1; APO-1; Apo1 antigen; Apo-1 antigen; APO-1 cell surface antigen; Apo-1 Fas; apoptosis antigen 1; apoptosis signaling receptor FAS; apoptosis-mediating surface antigen FAS; APT1; CD95; CD95 antigen; FAS; Fas (TNF receptor superfamily member 6); Fas (TNF receptor superfamily member); Fas (TNF receptor superfamily, member 6); Fas AMA; Fas antigen; Fas antigen (ATP1); Fas cell surface death receptor; Fas receptor; FAS1; FASLG receptor; FASTM; lpr; lymphoproliferation; OTTHUMP00000020045; OTTHUMP00000020046; OTTHUMP00000020051; OTTHUMP00000059646; TNF receptor superfamily member 6; TNFR6; Tnfrsf6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 26; Tumor necrosis factor receptor superfamily, member 6 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
DX2 | |
Flow Cytometry | |
NovaFluor Red 700 | |
Mouse | |
Human, Canine | |
P25445 | |
355, 486469 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
FAS |