missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 (NCAM) Monoclonal Antibody (TULY56), NovaFluor™ Yellow 690, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$604.08 - $1340.54
Specifications
Antigen | CD56 (NCAM) |
---|---|
Clone | TULY56 |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H013T02Y05A
|
Invitrogen™
H013T02Y05A |
25 Tests |
Each for $604.08
|
|
|||||
H013T03Y05A
|
Invitrogen™
H013T03Y05A |
100 Tests |
Each for $1,340.54
|
|
|||||
Description
CD56 (NCAM) Monoclonal antibody specifically detects CD56 (NCAM) in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow CytometrySpecifications
CD56 (NCAM) | |
0.2 μg/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
TULY56 | |
Flow Cytometry | |
NovaFluor Yellow 690 | |
Mouse | |
Human, Monkey, Rhesus Macaque | |
P13591 | |
4684, 693789 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Ncam1 |