missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RB Monoclonal Antibody (PD7/26), Alexa Fluor™ Plus 647
CD45RB Monoclonal Antibody (PD7/26), Alexa Fluor™ Plus 647
Supplier: Fisher Chemical 756945882
Description
CD45RB Monoclonal Antibody for IHC (P)
Specifications
CD45RB | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.5% BSA, 10% proprietary stabilizer and 0.05% sodium azide; pH 7.2 | |
P08575 | |
Ptprc | |
Affinity Chromatography | |
RUO | |
5788 | |
4°C, store in dark | |
Liquid |
Immunohistochemistry (Paraffin) | |
PD7/26 | |
Alexa Fluor Plus 647 | |
Ptprc | |
B220; CD45; CD45 antigen; CD45R; GP180; LCA; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |