missing translation for 'onlineSavingsMsg'
Learn More

CD40 Monoclonal Antibody (1C10), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

$661.79 - $1340.54

Specifications

Antigen CD40
Clone 1C10
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
37-604-0180
View Documents
Invitrogen™
376040180
25 μg
Each for $661.79
Only null left
 
37-604-0182
View Documents
Invitrogen™
376040182
100 μg
Each for $1,340.54
Only null left
 
Description

Description

CD40 Monoclonal antibody specifically detects CD40 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD40
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5
IgG2a κ
Affinity chromatography
Antibody
1C10
Flow Cytometry
Brilliant Ultraviolet 661
Rat
Mouse
P27512
21939
Primary
4°C, store in dark, DO NOT FREEZE!
CD40
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'