missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Monoclonal Antibody (GK1.5), NovaFluor™ Blue 800, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$604.08 - $1170.85
Specifications
Antigen | CD4 |
---|---|
Clone | GK1.5 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M001T03B12A
|
Fisher Chemical
M001T03B12A |
100 μg |
Each for $1,170.85
|
|
|||||
M001T02B12A
|
Fisher Chemical
M001T02B12A |
25 μg |
Each for $604.08
|
|
|||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow CytometrySpecifications
CD4 | |
0.1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
GK1.5 | |
Flow Cytometry | |
NovaFluor Blue 800 | |
Rat | |
Mouse | |
P06332 | |
12504 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD4 |