missing translation for 'onlineSavingsMsg'
Learn More

CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH1), NovaFluor™ Red 700, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$604.08 - $1340.54

Specifications

Antigen CD274 (PD-L1, B7-H1)
Clone MIH1
Concentration 0.6 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H089T02R03A
View Documents
Fisher Chemical
H089T02R03A
25 Tests
Each for $604.08
Only null left
 
H089T03R03A
View Documents
Fisher Chemical
H089T03R03A
100 Tests
Each for $1,340.54
Only null left
 
Description

Description

CD274 (PD-L1, B7-H1) Monoclonal antibody specifically detects CD274 (PD-L1, B7-H1) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD274 (PD-L1, B7-H1)
0.6 μg/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
IgG1 κ
Affinity chromatography
Antibody
MIH1
Flow Cytometry
NovaFluor Red 700
Mouse
Human
Q9NZQ7
29126
Primary
4° C, store in dark, DO NOT FREEZE!
Cd274
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'