missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH1), Brilliant Violet™ 786, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$607.48 - $1286.23
Specifications
Antigen | CD274 (PD-L1, B7-H1) |
---|---|
Clone | MIH1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
41-759-8342
|
Invitrogen™
417598342 |
100 Tests |
Each for $1,286.23
|
|
|||||
41-759-8341
|
Invitrogen™
417598341 |
25 Tests |
Each for $607.48
|
|
|||||
Description
CD274 (PD-L1, B7-H1) Monoclonal antibody specifically detects CD274 (PD-L1, B7-H1) in Human samples. It is validated for Flow CytometrySpecifications
CD274 (PD-L1, B7-H1) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
MIH1 | |
Flow Cytometry | |
Brilliant Violet 786 | |
Mouse | |
Human | |
Q9NZQ7 | |
29126 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Cd274 |