missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD268 (BAFF Receptor) Monoclonal Antibody (8A7), NovaFluor™ Blue 690, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ H078T02B09A
Description
Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13. 1-q13. 31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.Specifications
CD268 (BAFF Receptor) | |
Monoclonal | |
0.4 μg/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q96RJ3 | |
TNFRSF13C | |
Affinity chromatography | |
RUO | |
115650 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
8A7 | |
NovaFluor Blue 690 | |
TNFRSF13C | |
2010006P15Rik; B cell-activating factor receptor; BAFF receptor; BAFFR; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13C; Tnfrsf13c; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a κ |