missing translation for 'onlineSavingsMsg'
Learn More

CD25 Monoclonal Antibody (PC61.5), NovaFluor™ Blue 585, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

$604.08 - $1170.85

Specifications

Antigen CD25
Clone PC61.5
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M060T03B04A
View Documents
Invitrogen™
M060T03B04A
100 μg
Each for $1,170.85
Only null left
 
M060T02B04A
View Documents
Invitrogen™
M060T02B04A
25 μg
Each for $604.08
Only null left
 
Description

Description

CD25 Monoclonal antibody specifically detects CD25 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD25
0.1 mg/mL
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR
IgG1 λ
Affinity chromatography
Antibody
PC61.5
Flow Cytometry
NovaFluor Blue 585
Rat
Mouse
P01590
16184
Primary
4°C, store in dark, DO NOT FREEZE!
IL2RA
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'