missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Monoclonal Antibody (MAC 2-158), NovaFluor™ Red 725, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$604.08 - $1340.54
Specifications
Antigen | CD163 |
---|---|
Clone | MAC 2-158 |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H088T02R05A
|
Fisher Chemical
H088T02R05A |
25 Tests |
Each for $604.08
|
|
|||||
H088T03R05A
|
Fisher Chemical
H088T03R05A |
100 Tests |
Each for $1,340.54
|
|
|||||
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow CytometrySpecifications
CD163 | |
0.2 μg/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
MAC 2-158 | |
Flow Cytometry | |
NovaFluor Red 725 | |
Mouse | |
Human | |
Q86VB7 | |
9332 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD163 |