missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD OptiCHO™ AGT™ Medium DFS Item

CD OptiCHO™ AGT™ Medium

$205.00 - $13300.00

Specifications

Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Powder (AGT)
Product Type CD OptiCHO AGT Medium
Serum Level Serum-free
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
50-114-8223 DFS Item
missing translation for 'viewDocuments'
Gibco™
A1122204
1 L
Each for $205.00
Only null left
 
A1122201 DFS Item
missing translation for 'viewDocuments'
Gibco™
A1122201
100 L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

CD OptiCHO™ medium is specifically designed to offer high performance and yield with recombinant CHO cells in a chemically defined fed-batch environment. The protein-free, animal origin free formulation is designed to provide consistency and reduces the need to screen for adventitious agents. CD OptiCHO™ medium builds on Gibco™ CD CHO’s legacy of consistent cell growth and titers, while offering up to 40% better titer and cell growth over select competitors. Thanks to its low osmolality and advanced CHO CD EfficientFeed™ Liquid Nutrient Supplement Kit feeding options, CD OptiCHO™ medium is capable of gaining 2-5 fold increases in fed-batch productivity over batch culture.

When to Use CD OptiCHO™ Medium
  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is finicky, i.e. hard to grow or adapt

    When to Consider Another Gibco™ Medium

    Consider CD FortiCHO™ medium when:
  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

    Consider CD CHO medium when:
  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

    Consider CD DG44 medium when:
  • You use parental DG44 or DXB11 CHO cells (dhfr-)

    Liquid Media Formulations
    Gibco™ AGT™ media are also available in a liquid format that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.

    Design Your Own Media Specific to Your Needs
    Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
  • Specifications

    Specifications

    CHO
    Powder (AGT)
    Serum-free
    High Glucose, Sodium Pyruvate, Sodium Bicarbonate
    AGT™, Gibco™
    Low
    Magnesium, Sodium Phosphate
    CO2
    Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    CD OptiCHO AGT Medium
    Non-sterile
    No Glutamine, No Phenol Red
    Wet Ice
    No antibiotics
    Certified
    Suspension Cell Culture
    Videos
    SDS
    Documents

    Documents

    For Research Use Only. Not for use in diagnostic procedures.