Learn More
CritiCore CAT 1 Women's Lab Coat - Nomex™ FR
Lab Coat designed to provide protection against flash fire, electric arc (HRC 1), and other dangerous conditions. This Flame-Resistant lab coat is NFPA 2112:2023 Certified with numerous applications in both laboratory and production settings.
$337.49 - $472.52
Specifications
Product Type Flame-resistant Lab Coat
Gender Women's
Material Nomex
Color Blue
Closure Type Snaps
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17012877
View Documents
CritiCore
12L34950CXP63PSBLUE
Small
Each for $337.49
Only null left
17012878
View Documents
CritiCore
12L34950CXP63PMBLUE
Medium
Each for $337.49
Only null left
17012879
View Documents
CritiCore
12L34950CXP63PLBLUE
Large
Each for $337.49
Only null left
17012880
View Documents
CritiCore
12L34950CXP63PXLBLUE
X-Large
Each for $337.49
Only null left
17012881
View Documents
CritiCore
12L34950CXP63P2XLBLUE
2X-Large
Each for $371.24
Only null left
17012882
View Documents
CritiCore
12L34950CXP63P3XLBLUE
3X-Large
Each for $404.98
Only null left
17012883
View Documents
CritiCore
12L34950CXP63P4XLBLUE
4X-Large
Each for $438.76
Only null left
17012884
View Documents
CritiCore
12L34950CXP63P5XLBLUE
5X-Large
Each for $472.52
Only null left
Description
Description
  • Fabric: Nomex™ FR
  • Blend: 100% Nomex™ IIIA,: prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
  • Arc Rating (ATPV) - 6.5 cal/sq cm
  • Quick-release snap front
  • Safety sleeve cuff with tunneled elastic
  • FR identification patches on sleeves and back
  • Excellent heat-resistance properties
  • Includes a waistband in the back for a more tailored fit
  • NFPA 2112:2023 Certified
Specifications
Specifications
Flame-resistant Lab Coat
Nomex
Snaps
Reusable
Flame
3
Women's
Blue
Elastic
Non-sterile
Standard Length
Sewn
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'