missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose Sodium, High Viscosity, USP, Spectrum™ Chemical

CA194, 9004-32-4

$403.00 - $9676.83

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601471
View Documents
Spectrum Chemical Mfg Cor
CA194125GMPL
125 g Poly Bottle
Each for $403.00
Only null left
 
18601472
View Documents
Spectrum Chemical Mfg Cor
CA194500GMPL
500 g Poly Bottle
Each for $1,015.33
Only null left
 
18601473
View Documents
Spectrum Chemical Mfg Cor
CA19425KGPL
2.5 kg Poly Pail
Each for $2,647.73
Only null left
 
18601474
View Documents
Spectrum Chemical Mfg Cor
CA19412KG
12 kg Fiber Drum
Each for $9,676.83
Only null left
 
Description

Description

Spectrum™ Chemical Carboxymethylcellulose Sodium, High Viscosity, Viscosity of 1 Percent Aqueous Solution @ 25 DEG C: 1500-3000 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

9004-32-4
263.20
DPXJVFZANSGRMM-UHFFFAOYNA-N
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
(C12 H14 O9 R6)n
MFCD00081472
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium

Specifications

9004-32-4
(C12 H14 O9 R6)n
125 g
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
263.20
10%
100%
MFCD00081472
DPXJVFZANSGRMM-UHFFFAOYNA-N
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
USP
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'