missing translation for 'onlineSavingsMsg'
Learn More

Carbon Disulfide, Reagent Grade, ≥99.9%, Low benzene, Honeywell™

Reagent Grade, low benzene,≥99.9%

$234.40 - $805.35

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046321
View Documents
Honeywell Chemicals
342270100ML
100 mL Glass Bottle
Each for $270.48
Only null left
 
60046323
View Documents
Honeywell Chemicals
342270800ML
800 mL Glass Bottle
Each for $234.40
Only null left
 
60046322
View Documents
Honeywell Chemicals
3422701L
1 L Glass Bottle
Each for $805.35
Only null left
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

75-15-0
76.131
QGJOPFRUJISHPQ-UHFFFAOYSA-N
6348
methanedithione
CS2
MFCD00011321
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
CHEBI:23012
C(=S)=S

Specifications

≤-76°C
Colorless
46°C
0.5
212°F
Glass Bottle
CS2
UN1131
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
QGJOPFRUJISHPQ-UHFFFAOYSA-N
methanedithione
6348
76.14g/mol
ReagentPlus™
75-15-0
1.266g/mL at 25°C
100 mL
−30°C (−22°F)
Unpleasant
Liquid
MFCD00011321
1098293
2.9g/L at 20°C in water
C(=S)=S
76.131
CHEBI:23012
≥99.9%
Carbon disulfide, Low benzene
Videos
Safety and Handling

Safety and Handling

P201-P210-P261-P280-P304 + P340 + P312-P308 + P313

DOTInformation : class 3 - PG 1 - Carbon disulfide

RTECSNumber : 200-843-6

SDS
missing translation for 'documents'

missing translation for 'documents'