missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haen

CHROMASOLV™, for HPLC,≥99.9%

Supplier:  Honeywell Chemicals 2706604X2L

 View more versions of this product

Catalog No. 6000395


Only null left
Explore available promotions

Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Specifications

Carbon disulfide
CHROMASOLV™, for HPLC, ≥99.9%
max. 0.50 at 388 nm, max. 1.0 at 386 nm, max. 0.01 at 750 nm, max. 0.01 500 nm, max. 0.05 at 410 nm, max. 0.10 at 403 nm, max. 0.25 at 394 nm
1.266 g/mL at 25 °C(lit.)
4 x 2 L
Glass Bottle
CS2
UN1131
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
C(=S)=S
76.131
CHEBI:23012
>99.9%
-112°C to -111°C (literature)
75-15-0
100.0000%
46°C
−30°C
CS2
MFCD00011321
1098293
QGJOPFRUJISHPQ-UHFFFAOYSA-N
methanedithione
6348
76.14
HPLC

Chemical Identifiers

75-15-0
76.131
QGJOPFRUJISHPQ-UHFFFAOYSA-N
6348
methanedithione
CS2
MFCD00011321
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
CHEBI:23012
C(=S)=S
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 1260 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Carbon disulfide

EINECSNumber : 200-843-6

Recommended Storage : Room Temp

SDS
Documents

Documents

Promotions

Promotions