missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamazepin 97.0+%, TCI America™
$83.89 - $162.17
Chemical Identifiers
CAS | 298-46-4 |
---|---|
Molecular Formula | C15H12N2O |
Molecular Weight (g/mol) | 236.27 |
MDL Number | MFCD00005073 |
InChI Key | FFGPTBGBLSHEPO-UHFFFAOYSA-N |
Synonym | carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston |
PubChem CID | 2554 |
ChEBI | CHEBI:3387 |
IUPAC Name | 2-azatricyclo[9.4.0.0³,â¸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide |
SMILES | NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12 |
Chemical Identifiers
298-46-4 | |
236.27 | |
FFGPTBGBLSHEPO-UHFFFAOYSA-N | |
2554 | |
2-azatricyclo[9.4.0.0³,â¸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide |
C15H12N2O | |
MFCD00005073 | |
carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston | |
CHEBI:3387 | |
NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12 |
Specifications
298-46-4 | |
White | |
MFCD00005073 | |
carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston | |
NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12 | |
236.27 | |
CHEBI:3387 | |
≥97.0% (N) | |
Carbamazepin |
191°C | |
C15H12N2O | |
10 g | |
FFGPTBGBLSHEPO-UHFFFAOYSA-N | |
2-azatricyclo[9.4.0.0³,â¸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide | |
2554 | |
236.27 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : HN8225000
TSCA : No