missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Camphoric Acid 98.0+%, TCI America™
$156.59 - $430.60
Chemical Identifiers
CAS | 124-83-4 |
---|---|
Molecular Formula | C10H14O4-2 |
Molecular Weight (g/mol) | 198.218 |
MDL Number | MFCD00001375 |
InChI Key | LSPHULWDVZXLIL-LDWIPMOCSA-L |
Synonym | d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid |
PubChem CID | 6918944 |
IUPAC Name | (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate |
SMILES | CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C |
Chemical Identifiers
124-83-4 | |
198.218 | |
LSPHULWDVZXLIL-LDWIPMOCSA-L | |
6918944 | |
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C |
C10H14O4-2 | |
MFCD00001375 | |
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid | |
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate |
Specifications
187°C | |
White | |
MFCD00001375 | |
LSPHULWDVZXLIL-LDWIPMOCSA-L | |
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate | |
6918944 | |
≥98.0% (GC,T) | |
Crystalline Powder |
124-83-4 | |
C10H14O4-2 | |
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid | |
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C | |
198.218 | |
200.23 | |
25 g | |
(+)-Camphoric Acid |
Safety and Handling
EINECSNumber : (4)-0308
TSCA : Yes