missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain 8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567316
Description
Immunogen sequence: FDGFNINTCRE MISLLDSNGT GTLGAVEFKT LWLKIQKYLE IYWETDYNHS GTIDAHEMRT AL Highest antigen sequence identity to the following orthologs - mouse 81%, rat 81%.
Calcium-regulated non-lysosomal thiol-protease. Involved in membrane trafficking in the gastric surface mucus cells (pit cells) and may involve the membrane trafficking of mucus cells via interactions with coat protein. Proteolytically cleaves the beta- subunit of coatomer complex.Specifications
Calpain 8 | |
Polyclonal | |
Unconjugated | |
CAPN8 | |
calpain 8; calpain-8; CAPN8; NCL2; nCL-2; New calpain 2; stomach-specific M-type calpain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388743 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NHC0 | |
CAPN8 | |
Recombinant Human Calpain 8. Recombinant protein control fragment (Product #RP-107958). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |