missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, For BOD, Certified, 2.75% (w/v) ±0.05%, LabChem™
$53.07 - $76.24
Chemical Identifiers
10035-04-8 |
Specifications
Colorless | |
96.36,3.64 | |
Soluble in water | |
2.75% ±0.05% (w/v) | |
Passes Test | |
Traceable to NIST | |
500 mL | |
Calcium Chloride, For BOD |
10035-04-8,7732-18-5 | |
CaCl2·2H2O | |
147.02 | |
Certified | |
Poly Bottle | |
Hydrogen chloride | |
Liquid |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Refrigerate