missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cadmium phosphide, 99.5% (metals basis)

$70.45 - $522.53

Specifications

Quantity 1 g
Percent Purity 99.5%

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8721903
View Documents
Thermo Scientific™
8721903
1 g
Each for $70.45
Only null left
 
AA8721909
View Documents
Thermo Scientific™
8721909
10 g
Each for $522.53
Only null left
 
Description

Description

Specifications

Specifications

1 g
99.5%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'