missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cacotheline Monohydrate 98.0+%, TCI America™
$410.10 - $445.51
Chemical Identifiers
CAS | 561-20-6 |
---|---|
Molecular Formula | C21H21N3O7 |
Molecular Weight (g/mol) | 427.41 |
MDL Number | MFCD00077211 |
InChI Key | IVEMPCACOMNRGI-UHFFFAOYNA-N |
PubChem CID | 101667989 |
IUPAC Name | 2-{9-nitro-7,8-dioxo-15-oxa-1,11-diazahexacyclo[16.3.1.0â´,¹².0â´,²¹.0âµ,¹â°.0¹³,¹â¹]docosa-5,9,17-trien-14-yl}acetic acid |
SMILES | OC(=O)CC1OCC=C2CN3CCC45C3CC2C1C4NC1=C(C(=O)C(=O)C=C51)[N+]([O-])=O |
Chemical Identifiers
561-20-6 | |
427.41 | |
IVEMPCACOMNRGI-UHFFFAOYNA-N | |
2-{9-nitro-7,8-dioxo-15-oxa-1,11-diazahexacyclo[16.3.1.0â´,¹².0â´,²¹.0âµ,¹â°.0¹³,¹â¹]docosa-5,9,17-trien-14-yl}acetic acid |
C21H21N3O7 | |
MFCD00077211 | |
101667989 | |
OC(=O)CC1OCC=C2CN3CCC45C3CC2C1C4NC1=C(C(=O)C(=O)C=C51)[N+]([O-])=O |
Specifications
561-20-6 | |
MFCD00077211 | |
2811 | |
OC(=O)CC1OCC=C2CN3CCC45C3CC2C1C4NC1=C(C(=O)C(=O)C=C51)[N+]([O-])=O | |
427.41 | |
490.43 | |
Crystalline Powder |
C21H21N3O7 | |
1 g | |
IVEMPCACOMNRGI-UHFFFAOYNA-N | |
2-{9-nitro-7,8-dioxo-15-oxa-1,11-diazahexacyclo[16.3.1.0â´,¹².0â´,²¹.0âµ,¹â°.0¹³,¹â¹]docosa-5,9,17-trien-14-yl}acetic acid | |
101667989 | |
≥98.0% (T) | |
Cacotheline Monohydrate |
Safety and Handling
TSCA : Yes