missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyric Acid 99.0+%, TCI America™
$29.83 - $54.06
Chemical Identifiers
CAS | 107-92-6 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.11 |
MDL Number | MFCD00002814 |
InChI Key | FERIUCNNQQJTOY-UHFFFAOYSA-N |
Synonym | butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure |
PubChem CID | 264 |
ChEBI | CHEBI:30772 |
IUPAC Name | butanoic acid |
SMILES | CCCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B075425ML
|
TCI America
B075425ML |
25 mL |
Each for $29.83
|
|
|||||
B0754500ML
|
TCI America
B0754500ML |
500 mL |
Each for $54.06
|
|
|||||
Chemical Identifiers
107-92-6 | |
88.11 | |
FERIUCNNQQJTOY-UHFFFAOYSA-N | |
264 | |
butanoic acid |
C4H8O2 | |
MFCD00002814 | |
butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure | |
CHEBI:30772 | |
CCCC(O)=O |
Specifications
107-92-6 | |
Colorless | |
MFCD00002814 | |
butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure | |
CCCC(O)=O | |
88.11 | |
CHEBI:30772 | |
≥99.0% (GC,T) | |
Liquid |
163°C | |
C4H8O2 | |
2820 | |
FERIUCNNQQJTOY-UHFFFAOYSA-N | |
butanoic acid | |
264 | |
88.11 | |
25 mL | |
Butyric Acid |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : ES5425000
TSCA : Yes