missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butylamine Hydrochloride 98.0+%, TCI America™
$76.43 - $452.97
Chemical Identifiers
CAS | 3858-78-4 |
---|---|
Molecular Formula | C4H12ClN |
Molecular Weight (g/mol) | 109.597 |
MDL Number | MFCD00043791 |
InChI Key | ICXXXLGATNSZAV-UHFFFAOYSA-N |
Synonym | 1-Aminobutane Hydrochloride, Butylammonium Chloride |
PubChem CID | 6432400 |
IUPAC Name | butan-1-amine;hydrochloride |
SMILES | CCCCN.Cl |
Chemical Identifiers
3858-78-4 | |
109.597 | |
ICXXXLGATNSZAV-UHFFFAOYSA-N | |
6432400 | |
CCCCN.Cl |
C4H12ClN | |
MFCD00043791 | |
1-Aminobutane Hydrochloride, Butylammonium Chloride | |
butan-1-amine;hydrochloride |
Specifications
3858-78-4 | |
White | |
MFCD00043791 | |
1-Aminobutane Hydrochloride, Butylammonium Chloride | |
CCCCN.Cl | |
109.597 | |
109.60 | |
Crystalline Powder |
214°C | |
C4H12ClN | |
25 g | |
ICXXXLGATNSZAV-UHFFFAOYSA-N | |
butan-1-amine;hydrochloride | |
6432400 | |
≥98.0% (T) | |
Butylamine Hydrochloride |
Safety and Handling
EINECSNumber : (2)-0132
RTECSNumber : EO5199990
TSCA : Yes